This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01113688_m1
See other DLGAP2 GE Assays ›
Gene Symbol:
DLGAP2
Gene Name:
DLG associated protein 2
Gene Aliases:
DAP2, SAPAP2
Chromosome Location:
Chr.8: 1501366 - 1708476 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01113688_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLGAP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277161.1 NP_001264090.1 1 - 2 73 93
NM_004745.4 NP_004736.2 1 - 2 73 93
GenBank mRNA AF119818.1 - 1 - 2 73 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001277161.1, NM_004745.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DLGAP2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

neuron-neuron synaptic transmission
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0