This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01113252_g1
See other TRA2A GE Assays ›
Gene Symbol:
TRA2A
Gene Name:
transformer 2 alpha homolog
Gene Aliases:
AWMS1, HSU53209
Chromosome Location:
Chr.7: 23504780 - 23532041 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01113252_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRA2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282757.1 NP_001269686.1 4 - 5 857 97
NM_001282758.1 NP_001269687.1 4 - 5 646 97
NM_001282759.1 NP_001269688.1 5 - 6 947 97
NM_013293.4 NP_037425.1 3 - 4 556 97
XM_005249725.1 XP_005249782.1 4 - 5 843 97
XM_017012089.1 XP_016867578.1 3 - 4 1088 97
XM_017012090.1 XP_016867579.1 5 - 6 898 97
GenBank mRNA AB097048.1 - 3 - 4 497 97
AK298815.1 - 5 - 6 908 97
AK300565.1 - 4 - 5 799 97
AK300741.1 - 4 - 5 586 97
AK310719.1 - 3 - 4 1088 97
AK311687.1 - 4 - 5 808 97
AK314801.1 - 3 - 4 497 97
BC017094.2 - 3 - 4 467 3907327 97
BG722500.1 - 3 - 4 560 97
U53209.1 - 3 - 4 496 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK298815.1, AK300565.1, AK300741.1, NM_001282757.1, NM_001282758.1, NM_001282759.1, XM_005249725.1, XM_017012089.1, XM_017012090.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRA2A

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
nucleotide binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0