This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01112760_g1
See other NAV3 GE Assays ›
Gene Symbol:
NAV3
Gene Name:
neuron navigator 3
Gene Aliases:
POMFIL1, STEERIN3, unc53H3
Chromosome Location:
Chr.12: 77571862 - 78213012 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01112760_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAV3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024383.1 NP_001019554.1 27 - 28 5474 70
XM_017020164.1 XP_016875653.1 26 - 27 9456 70
XM_017020165.1 XP_016875654.1 25 - 26 9414 70
XM_017020166.1 XP_016875655.1 27 - 28 5463 70
XM_017020167.1 XP_016875656.1 26 - 27 5394 70
XM_017020168.1 XP_016875657.1 26 - 27 8987 70
XM_017020169.1 XP_016875658.1 25 - 26 8944 70
XM_017020171.1 XP_016875660.1 19 - 20 8770 70
XM_017020173.1 XP_016875662.1 18 - 19 8279 70
XM_017020174.1 XP_016875663.1 17 - 18 8237 70
GenBank mRNA AJ488201.1 - 27 - 28 5346 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200008269.hg.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Process(es)

ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1200160743.hg.1 ENST00000397909.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200160743.hg.1 NM_001024383.1 Refseq
Affymetrix Human Clariom D Assay PSR1200160743.hg.1 ENST00000397909.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200160743.hg.1 NM_001024383.1 Refseq
Affymetrix Human Clariom D Assay PSR1200160743.hg.1 TC1200008269.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0