This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01111208_m1
See other MTMR6 GE Assays ›
Gene Symbol:
MTMR6
Gene Name:
myotubularin related protein 6
Gene Aliases:
-
Chromosome Location:
Chr.13: 25246201 - 25287566 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01111208_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTMR6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004685.3 NP_004676.3 6 - 7 1045 74
XM_017020845.1 XP_016876334.1 6 - 7 991 74
GenBank mRNA AF072928.1 - 2 - 3 259 74
AF406619.1 - 6 - 7 726 74
AK314587.1 - 6 - 7 910 74
AL832017.2 - 6 - 7 968 74
BC040012.1 - 6 - 7 917 5495564 74
BU177677.1 - 1 - 2 110 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008386.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000498.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MTMR6

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
phosphatidylinositol dephosphorylation
potassium ion transmembrane transport
phosphatidylinositol-3-phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
calcium-activated potassium channel activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 ENST00000319298.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 ENST00000540661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 ENST00000541021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005645.hg.1 TC13000498.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156608.hg.1 TC1300008386.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0