This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01111203_m1
See other MTMR6 GE Assays ›
Gene Symbol:
MTMR6
Gene Name:
myotubularin related protein 6
Gene Aliases:
-
Chromosome Location:
Chr.13: 25246201 - 25287566 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01111203_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTMR6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004685.3 NP_004676.3 1 - 2 345 71
XM_011535306.1 XP_011533608.1 1 - 2 291 71
XM_011535307.1 XP_011533609.1 1 - 2 291 71
XM_017020845.1 XP_016876334.1 1 - 2 291 71
GenBank mRNA AK314587.1 - 1 - 2 210 71
BC040012.1 - 1 - 2 217 5495564 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008386.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000498.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK314587.1,BC040012.1,NM_004685.3,XM_011535306.1,XM_011535307.1,XM_017020845.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MTMR6

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
phosphatidylinositol dephosphorylation
potassium ion transmembrane transport
phosphatidylinositol-3-phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
calcium-activated potassium channel activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005652.hg.1 ENST00000319298.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005652.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005652.hg.1 ENST00000540661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005652.hg.1 ENST00000541021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005652.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005652.hg.1 TC13000498.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13005654.hg.1 ENST00000319298.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005654.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005654.hg.1 ENST00000540661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005654.hg.1 ENST00000541021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005654.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005654.hg.1 TC13000498.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13005653.hg.1 ENST00000319298.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005653.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005653.hg.1 ENST00000540661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005653.hg.1 ENST00000541021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005653.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005653.hg.1 TC13000498.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13005655.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005655.hg.1 ENST00000540661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005655.hg.1 ENST00000541021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005655.hg.1 NM_004685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005655.hg.1 TC13000498.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300156618.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156618.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156618.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156618.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156618.hg.1 TC1300008386.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300156615.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156615.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156615.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156615.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156615.hg.1 TC1300008386.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300156617.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156617.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156617.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300156617.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay PSR1300156617.hg.1 TC1300008386.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000381801.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 ENST00000482345.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq
Affymetrix Human Clariom D Assay TC1300008386.hg.1 NM_004685.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0