Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001100.hg.1 |
ENST00000267067.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001100.hg.1 |
ENST00000380235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001100.hg.1 |
ENST00000380250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001100.hg.1 |
ENST00000380257.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001100.hg.1 |
NM_023037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001100.hg.1 |
TC13000109.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
ENST00000380250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
ENST00000542859.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
NM_023037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
ENST00000380250.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
ENST00000542859.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
NM_023037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146226.hg.1 |
TC1300006799.hg.1 |
Clariom D TC |