This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01110981_m1
See other GUCY2EP GE Assays ›
Gene Symbol:
GUCY2EP
Gene Name:
guanylate cyclase 2E, pseudogene
Gene Aliases:
GC-E, GCD, GUCY2E
Chromosome Location:
Chr.11: 76680166 - 76721789 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01110981_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GUCY2EP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024042.1 17 - 18 2288 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream leucine rich repeat containing 32 LRRC32 D11S833E,
GARP
-20.7kb Sense Chr. 11 - 76657441 - 76670941 on NCBI Build GRCh38
Downstream tsukushi,
small leucine rich proteoglycan
TSKU E2IG4,
LRRC54,
TSK
88.2kb Anti-sense Chr. 11 - 76782313 - 76798154 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011695.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002117.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11025269.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11025269.hg.1 TC11002117.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11025270.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11025270.hg.1 TC11002117.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100190228.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190228.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190228.hg.1 TC1100011695.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100190227.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190227.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190227.hg.1 TC1100011695.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0