This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01110978_m1
See other GUCY2EP GE Assays ›
Gene Symbol:
GUCY2EP
Gene Name:
guanylate cyclase 2E, pseudogene
Gene Aliases:
GC-E, GCD, GUCY2E
Chromosome Location:
Chr.11: 76680166 - 76721789 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01110978_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GUCY2EP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024042.1 14 - 15 1901 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream leucine rich repeat containing 32 LRRC32 D11S833E,
GARP
-24.4kb Sense Chr. 11 - 76657441 - 76670941 on NCBI Build GRCh38
Downstream tsukushi,
small leucine rich proteoglycan
TSKU E2IG4,
LRRC54,
TSK
85.4kb Anti-sense Chr. 11 - 76782313 - 76798154 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011695.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002117.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11025273.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11025273.hg.1 TC11002117.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190234.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100190234.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190234.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100190234.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190234.hg.1 TC1100011695.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000525328.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000526984.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 ENST00000531117.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay TC1100011695.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190235.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190235.hg.1 NR_024042.1 Refseq
Affymetrix Human Clariom D Assay PSR1100190235.hg.1 TC1100011695.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0