This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01110896_m1
See other HSPA14 GE Assays ›
Gene Symbol:
HSPA14
Gene Name:
heat shock protein family A (Hsp70) member 14
Gene Aliases:
HSP70-4, HSP70L1
Chromosome Location:
Chr.10: 14838160 - 14871741 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01110896_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSPA14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_103464.1 3 - 4 468 109
GenBank mRNA AK000880.1 - 3 - 4 309 109
BC065281.1 - 3 - 4 299 109
BC108738.1 - 3 - 4 402 109
BQ424016.1 - 3 - 4 399 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000006858.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK000880.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

'de novo' cotranslational protein folding
protein binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay PSR1000146929.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000146929.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay PSR1000146929.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000146929.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay PSR1000146929.hg.1 TC1000006858.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay PSR1000146928.hg.1 TC1000006858.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0