This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01108618_m1
See other SMCHD1 GE Assays ›
Gene Symbol:
SMCHD1
Gene Name:
structural maintenance of chromosomes flexible hinge domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 2655887 - 2805017 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01108618_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMCHD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015295.2 NP_056110.2 40 - 41 5242 165
XM_011525642.1 XP_011523944.1 40 - 41 5242 165
XM_011525643.2 XP_011523945.1 40 - 41 5242 165
XM_011525644.2 XP_011523946.1 39 - 40 5150 165
XM_017025684.1 XP_016881173.1 41 - 42 5420 165
GenBank mRNA AB014550.1 - 14 - 15 1584 165
AL080138.1 - 11 - 12 1222 165
CR627458.1 - 29 - 30 3466 165

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800006487.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000011.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromosome organization
inactivation of X chromosome by DNA methylation
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR18000145.hg.1 ENST00000261598.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18000145.hg.1 ENST00000320876.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18000145.hg.1 NM_015295.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18000145.hg.1 TC18000011.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 ENST00000320876.10 Ensembl
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 ENST00000577880.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 ENST00000584897.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 NM_015295.2 Refseq
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 ENST00000320876.10 Ensembl
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 ENST00000577880.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 ENST00000584897.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 NM_015295.2 Refseq
Affymetrix Human Clariom D Assay PSR1800144467.hg.1 TC1800006487.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0