This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01107469_m1
See other ACOXL GE Assays ›
Gene Symbol:
ACOXL
Gene Name:
acyl-CoA oxidase-like
Gene Aliases:
-
Chromosome Location:
Chr.2: 110732539 - 111118571 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01107469_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACOXL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011511411.2 XP_011509713.1 9 - 10 1009 78
XM_017004431.1 XP_016859920.1 9 - 10 1009 78
GenBank mRNA BC022268.1 - 9 - 10 914 78
BX282060.1 - 1 - 2 75 78

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
fatty acid beta-oxidation using acyl-CoA dehydrogenase
fatty acid beta-oxidation using acyl-CoA oxidase
lipid homeostasis
fatty-acyl-CoA binding
molecular_function
acyl-CoA dehydrogenase activity
electron carrier activity
pristanoyl-CoA oxidase activity
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header