This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01107132_gH
See other SLC23A1 GE Assays ›
Gene Symbol:
SLC23A1
Gene Name:
solute carrier family 23 member 1
Gene Aliases:
SLC23A2, SVCT1, YSPL3
Chromosome Location:
Chr.5: 139367196 - 139384553 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01107132_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC23A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005847.4 NP_005838.3 10 - 11 1276 82
NM_152685.3 NP_689898.2 10 - 11 1288 82
XM_005272148.3 XP_005272205.3 11 - 12 1413 82
XM_005272149.3 XP_005272206.3 11 - 12 1401 82
XM_006714741.2 XP_006714804.2 11 - 12 1413 82
XM_011543765.2 XP_011542067.1 11 - 12 1413 82
XM_011543766.1 XP_011542068.1 10 - 11 1194 82
XM_011543767.1 XP_011542069.1 9 - 10 1098 82
XM_011543768.1 XP_011542070.1 10 - 11 1318 82
XM_011543769.1 XP_011542071.1 9 - 10 1073 82
GenBank mRNA AF058317.1 - 10 - 11 1179 82
AF170911.1 - 10 - 11 1210 82
AJ250807.1 - 10 - 11 1249 82
AJ269477.1 - 10 - 11 1179 82
AK092655.1 - 9 - 10 1100 82
AK290789.1 - 10 - 11 1209 82
AK298337.1 - 9 - 10 1109 82
BC050261.1 - 10 - 11 1230 5188430 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012204.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001840.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC23A1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
sodium ion transport
brain development
response to toxic substance
nucleobase transport
L-ascorbic acid transport
L-ascorbic acid metabolic process
lung development
vitamin transmembrane transport
sodium ion transmembrane transport
dehydroascorbic acid transport
transepithelial L-ascorbic acid transport
protein binding
L-ascorbate:sodium symporter activity
sodium ion transmembrane transporter activity
nucleobase transmembrane transporter activity
L-ascorbic acid transporter activity
dehydroascorbic acid transporter activity
sodium-dependent L-ascorbate transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000502863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000502863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000502863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025169.hg.1 ENST00000348729.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05025169.hg.1 ENST00000353963.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05025169.hg.1 ENST00000453898.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05025169.hg.1 NM_005847.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025169.hg.1 NM_152685.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025169.hg.1 TC05001840.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay PSR0500184616.hg.1 TC0500012204.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000348729.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000353963.7 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000502863.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000502863.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000502863.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000503919.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000504513.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000506512.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 ENST00000508270.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_005847.4 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq
Affymetrix Human Clariom D Assay TC0500012204.hg.1 NM_152685.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0