This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104673_m1
See other MAP4K3 GE Assays ›
Gene Symbol:
MAP4K3
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases:
GLK, MAPKKKK3, MEKKK 3, MEKKK3, RAB8IPL1
Chromosome Location:
Chr.2: 39249266 - 39437312 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104673_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAP4K3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003618.3 NP_003609.2 15 - 16 1443 104
GenBank mRNA AK291467.1 - 15 - 16 1409 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012320.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001765.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MAP4K3

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKKK activity
protein phosphorylation
JNK cascade
response to UV
response to tumor necrosis factor
intracellular signal transduction
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
MAP kinase kinase kinase kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02028465.hg.1 ENST00000263881.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02028465.hg.1 ENST00000341681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02028465.hg.1 ENST00000437545.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02028465.hg.1 ENST00000536018.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02028465.hg.1 NM_003618.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02028465.hg.1 TC02001765.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000263881.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000341681.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000414968.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000437545.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000465701.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000497566.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 NM_001270425.1 Refseq
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 NM_003618.3 Refseq
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000263881.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000341681.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000414968.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000437545.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000465701.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 ENST00000497566.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 NM_001270425.1 Refseq
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 NM_003618.3 Refseq
Affymetrix Human Clariom D Assay PSR0200190074.hg.1 TC0200012320.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0