This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104534_m1
See other SNTB2 GE Assays ›
Gene Symbol:
SNTB2
Gene Name:
syntrophin beta 2
Gene Aliases:
D16S2531E, EST25263, SNT2B2, SNT3, SNTL
Chromosome Location:
Chr.16: 69186133 - 69309052 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104534_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNTB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006750.3 NP_006741.1 5 - 6 1365 61
GenBank mRNA AF243385.1 - 4 - 5 1154 61
BC048215.1 - 5 - 6 1350 5805707 61
U40572.1 - 5 - 6 1365 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011420.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000565.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SNTB2

Gene Ontology Categories:

Process(es)

actin binding
structural molecule activity
protein binding
calmodulin binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16008170.hg.1 ENST00000336278.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16008170.hg.1 ENST00000467311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16008170.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16008170.hg.1 TC16000565.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191149.hg.1 TC1600011420.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0