This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104532_m1
See other SNTB2 GE Assays ›
Gene Symbol:
SNTB2
Gene Name:
syntrophin beta 2
Gene Aliases:
D16S2531E, EST25263, SNT2B2, SNT3, SNTL
Chromosome Location:
Chr.16: 69186133 - 69309052 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104532_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNTB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006750.3 NP_006741.1 2 - 3 812 75
GenBank mRNA BC048215.1 - 2 - 3 797 5805707 75
U40572.1 - 2 - 3 812 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011420.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000565.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SNTB2

Gene Ontology Categories:

Process(es)

actin binding
structural molecule activity
protein binding
calmodulin binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16008167.hg.1 ENST00000336278.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16008167.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16008167.hg.1 TC16000565.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16008166.hg.1 ENST00000336278.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16008166.hg.1 ENST00000524887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16008166.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16008166.hg.1 TC16000565.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000467311.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 ENST00000562422.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay TC1600011420.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000360496.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000524887.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191140.hg.1 TC1600011420.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600191143.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191143.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191143.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191143.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191143.hg.1 TC1600011420.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 ENST00000528525.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191144.hg.1 TC1600011420.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 ENST00000336278.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 ENST00000525632.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 NM_006750.3 Refseq
Affymetrix Human Clariom D Assay PSR1600191142.hg.1 TC1600011420.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0