This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104371_m1
See other ABHD5 GE Assays ›
Gene Symbol:
ABHD5
Gene Name:
abhydrolase domain containing 5
Gene Aliases:
CDS, CGI58, IECN2, NCIE2
Chromosome Location:
Chr.3: 43690883 - 43722725 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104371_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABHD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016006.4 NP_057090.2 2 - 3 245 148
XM_011533779.2 XP_011532081.1 2 - 3 495 148
XM_011533780.1 XP_011532082.1 2 - 3 192 148
GenBank mRNA AF151816.1 - 2 - 3 141 148
AK313811.1 - 2 - 3 155 148
BC021958.1 - 2 - 3 173 3920855 148
BG721387.1 - 2 - 3 264 148
KM659022.1 - 2 - 3 135 148
KU178625.1 - 2 - 3 135 148
KU178626.1 - 2 - 3 135 148

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300007194.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000231.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_011533779.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABHD5

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
negative regulation of sequestering of triglyceride
positive regulation of triglyceride catabolic process
cell differentiation
positive regulation of lipoprotein lipase activity
1-acylglycerol-3-phosphate O-acyltransferase activity
triglyceride lipase activity
lysophosphatidic acid acyltransferase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03004675.hg.1 ENST00000458276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004675.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004675.hg.1 TC03000231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004676.hg.1 ENST00000458276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004676.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004676.hg.1 TC03000231.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150706.hg.1 TC0300007194.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150707.hg.1 TC0300007194.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0