This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104370_m1
See other ABHD5 GE Assays ›
Gene Symbol:
ABHD5
Gene Name:
abhydrolase domain containing 5
Gene Aliases:
CDS, CGI58, IECN2, NCIE2
Chromosome Location:
Chr.3: 43690883 - 43722725 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104370_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABHD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016006.4 NP_057090.2 1 - 2 155 87
XM_011533780.1 XP_011532082.1 1 - 2 102 87
GenBank mRNA AF151816.1 - 1 - 2 51 87
AK313811.1 - 1 - 2 65 87
BC021958.1 - 1 - 2 83 3920855 87
BG721387.1 - 1 - 2 174 87
KM659022.1 - 1 - 2 45 87
KU178625.1 - 1 - 2 45 87
KU178626.1 - 1 - 2 45 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300007194.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000231.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABHD5

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
negative regulation of sequestering of triglyceride
positive regulation of triglyceride catabolic process
cell differentiation
positive regulation of lipoprotein lipase activity
1-acylglycerol-3-phosphate O-acyltransferase activity
triglyceride lipase activity
lysophosphatidic acid acyltransferase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03004670.hg.1 ENST00000458276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004670.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004670.hg.1 TC03000231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03004668.hg.1 ENST00000458276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004668.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004668.hg.1 TC03000231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004666.hg.1 ENST00000458276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004666.hg.1 NM_016006.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004666.hg.1 TC03000231.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150696.hg.1 TC0300007194.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000413300.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000463153.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay TC0300007194.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000454293.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000456453.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150701.hg.1 TC0300007194.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150698.hg.1 TC0300007194.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 ENST00000013894.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 ENST00000458276.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 ENST00000486764.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 NM_016006.5 Refseq
Affymetrix Human Clariom D Assay PSR0300150699.hg.1 TC0300007194.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0