This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01103426_m1
See other CMTR1 GE Assays ›
Gene Symbol:
CMTR1
Gene Name:
cap methyltransferase 1
Gene Aliases:
FTSJD2, KIAA0082, MTr1, hMTr1
Chromosome Location:
Chr.6: 37433131 - 37482844 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01103426_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CMTR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015050.2 NP_055865.1 4 - 5 700 86
XM_005248955.3 XP_005249012.1 4 - 5 630 86
GenBank mRNA AK292564.1 - 4 - 5 554 86
BC031890.1 - 4 - 5 594 4944457 86
D43949.2 - 4 - 5 590 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600007869.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000545.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CMTR1

Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
mRNA methylation
cap1 mRNA methylation
nucleic acid binding
mRNA (nucleoside-2'-O-)-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06005997.hg.1 ENST00000373427.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06005997.hg.1 ENST00000373451.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06005997.hg.1 NM_015050.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06005997.hg.1 TC06000545.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000457419.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000471097.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000475364.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 ENST00000493656.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay TC0600007869.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 ENST00000373451.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 ENST00000455891.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 NM_015050.2 Refseq
Affymetrix Human Clariom D Assay PSR0600154701.hg.1 TC0600007869.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0