Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008513.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008513.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008513.hg.1 |
ENST00000539286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008513.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008513.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008513.hg.1 |
TC07000584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008516.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008516.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008516.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008516.hg.1 |
TC07000584.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013405.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000474291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
ENST00000539286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
NM_001159491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197756.hg.1 |
TC0700013405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
ENST00000297290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
ENST00000473967.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
NM_015379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197759.hg.1 |
TC0700013405.hg.1 |
Clariom D TC |