Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001632.hg.1 |
ENST00000379059.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001632.hg.1 |
ENST00000379068.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001632.hg.1 |
NM_016937.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001632.hg.1 |
TC0X000120.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000379059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000379068.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000480125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000611764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
NM_016937.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000379059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000379068.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000480125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
ENST00000611764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
NM_016937.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146741.hg.1 |
TC0X00006823.hg.1 |
Clariom D TC |