This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01102323_g1
See other DPCD GE Assays ›
Gene Symbol:
DPCD
Gene Name:
deleted in primary ciliary dyskinesia homolog (mouse)
Gene Aliases:
-
Chromosome Location:
Chr.10: 101588332 - 101609653 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01102323_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DPCD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015448.1 NP_056263.1 3 - 4 271 72
GenBank mRNA AK290154.1 - 3 - 4 272 72
AY532267.1 - 3 - 4 258 72
BC001082.2 - 3 - 4 271 2907119 72
BC031695.1 - 3 - 4 294 5166605 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008686.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000739.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DPCD

Gene Ontology Categories:

Function(s) Process(es)

epithelial cilium movement
spermatogenesis
determination of left/right symmetry
lateral ventricle development
third ventricle development
sperm motility
left/right pattern formation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10008745.hg.1 ENST00000370147.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008745.hg.1 ENST00000370148.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008745.hg.1 ENST00000370149.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008745.hg.1 ENST00000370151.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10008745.hg.1 NM_015448.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10008745.hg.1 TC10000739.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000470165.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay TC1000008686.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000370147.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000370151.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000434727.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000475443.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 ENST00000626968.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 NM_015448.2 Refseq
Affymetrix Human Clariom D Assay PSR1000159399.hg.1 TC1000008686.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0