This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01102300_g1
See other HSPA14 GE Assays ›
Gene Symbol:
HSPA14
Gene Name:
heat shock protein family A (Hsp70) member 14
Gene Aliases:
HSP70-4, HSP70L1
Chromosome Location:
Chr.10: 14838160 - 14871741 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01102300_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSPA14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016299.3 NP_057383.2 4 - 5 505 128
GenBank mRNA AF112210.1 - 4 - 5 324 128
AF143723.1 - 4 - 5 314 128
AK225265.1 - 4 - 5 349 128
AK225651.1 - 4 - 5 349 128
AK292323.1 - 4 - 5 313 128
AK302452.1 - 4 - 5 349 128
BC119690.1 - 4 - 5 324 40116697 128
CN308284.1 - 4 - 5 342 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000006858.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000112.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HSPA14

Gene Ontology Categories:

Function(s) Process(es)

'de novo' cotranslational protein folding
protein binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10001221.hg.1 ENST00000378372.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10001221.hg.1 NM_016299.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10001221.hg.1 TC10000112.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000437161.6 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000470430.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493178.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 ENST00000493863.5 Ensembl
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_001278205.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay TC1000006858.hg.1 NR_103464.1 Refseq
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 ENST00000378372.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 ENST00000441647.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 NM_016299.3 Refseq
Affymetrix Human Clariom D Assay PSR1000146935.hg.1 TC1000006858.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0