Assay Details
Gene Symbol: GPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257199.1 | NP_001244128.1 | 6 - 7 | 1014 | 59 | |
NM_005276.3 | NP_005267.2 | 6 - 7 | 1083 | 59 | ||
GenBank mRNA | AK130162.1 | - | 6 - 7 | 825 | 59 | |
AK312516.1 | - | 6 - 7 | 865 | 59 | ||
BC032234.1 | - | 6 - 7 | 884 | 5229925 | 59 | |
L34041.1 | - | 6 - 7 | 880 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000388.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
gluconeogenesis
glycerophosphate shuttle
phosphatidic acid biosynthetic process
triglyceride biosynthetic process
positive regulation of glycolytic process
glycerol-3-phosphate catabolic process
cellular response to cAMP
cellular response to tumor necrosis factor
glycerol-3-phosphate dehydrogenase [NAD+] activity
glycerol-3-phosphate dehydrogenase activity
protein homodimerization activity
NAD binding
gluconeogenesis
glycerophosphate shuttle
phosphatidic acid biosynthetic process
triglyceride biosynthetic process
positive regulation of glycolytic process
glycerol-3-phosphate catabolic process
cellular response to cAMP
cellular response to tumor necrosis factor
glycerol-3-phosphate dehydrogenase [NAD+] activity
glycerol-3-phosphate dehydrogenase activity
protein homodimerization activity
NAD binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12004862.hg.1 | ENST00000301149.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12004862.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12004862.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12004862.hg.1 | TC12000388.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12004861.hg.1 | ENST00000301149.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12004861.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12004861.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12004861.hg.1 | TC12000388.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000547964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000550172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | ENST00000551939.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007596.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | ENST00000547190.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153779.hg.1 | TC1200007596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | ENST00000301149.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | ENST00000548814.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | NM_001257199.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | NM_005276.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153778.hg.1 | TC1200007596.hg.1 | Clariom D TC |
