This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01098803_m1
See other VWA8 GE Assays ›
Gene Symbol:
VWA8
Gene Name:
von Willebrand factor A domain containing 8
Gene Aliases:
KIAA0564
Chromosome Location:
Chr.13: 41566825 - 41961111 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01098803_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VWA8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009814.1 NP_001009814.1 20 - 21 2420 147
NM_015058.1 NP_055873.1 20 - 21 2420 147
XM_006719791.3 XP_006719854.1 12 - 13 1376 147
XM_011535007.2 XP_011533309.1 20 - 21 2446 147
XM_017020469.1 XP_016875958.1 22 - 23 2479 147
XM_017020470.1 XP_016875959.1 20 - 21 2446 147
XM_017020471.1 XP_016875960.1 20 - 21 2446 147
XM_017020472.1 XP_016875961.1 4 - 5 408 147
XM_017020474.1 XP_016875963.1 20 - 21 2446 147
GenBank mRNA AB011136.1 - 9 - 10 960 147
AK095155.1 - 20 - 21 2294 147
AL834181.1 - 13 - 14 1469 147
BC053674.1 - 20 - 21 2420 6171968 147
BC131802.1 - 21 - 22 2253 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008735.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000607.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VWA8

Gene Ontology Categories:

Process(es)

ATP binding
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13006930.hg.1 ENST00000251030.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13006930.hg.1 ENST00000281496.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13006930.hg.1 ENST00000379310.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13006930.hg.1 NM_001009814.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13006930.hg.1 NM_015058.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13006930.hg.1 TC13000607.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 ENST00000281496.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 ENST00000379310.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 NM_001009814.1 Refseq
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 NM_015058.1 Refseq
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 ENST00000281496.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 ENST00000379310.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 NM_001009814.1 Refseq
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 NM_015058.1 Refseq
Affymetrix Human Clariom D Assay PSR1300158944.hg.1 TC1300008735.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0