Assay Details
Gene Symbol: RBM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134363.2 | NP_001127835.2 | 10 - 11 | 2715 | 65 | |
XM_011539697.2 | XP_011537999.1 | 10 - 11 | 2553 | 65 | ||
XM_017016103.1 | XP_016871592.1 | 10 - 11 | 3973 | 65 | ||
XM_017016104.1 | XP_016871593.1 | 10 - 11 | 6189 | 65 | ||
GenBank mRNA | EU822950.1 | - | 10 - 11 | 2715 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000804.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10009640.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009640.hg.1 | ENST00000539821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009640.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009640.hg.1 | TC10000804.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10009641.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009641.hg.1 | ENST00000539821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009641.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009641.hg.1 | TC10000804.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000465774.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000471172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | ENST00000480343.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008898.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160845.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160845.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160845.hg.1 | ENST00000369519.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160845.hg.1 | NM_001134363.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160845.hg.1 | TC1000008898.hg.1 | Clariom D TC |
