This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01097167_g1
See other ATP6V1A GE Assays ›
Gene Symbol:
ATP6V1A
Gene Name:
ATPase H+ transporting V1 subunit A
Gene Aliases:
ATP6A1, ATP6V1A1, HO68, VA68, VPP2, Vma1
Chromosome Location:
Chr.3: 113747019 - 113812058 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01097167_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP6V1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001690.3 NP_001681.2 4 - 5 535 63
GenBank mRNA AF113129.1 - 4 - 5 462 63
AK293804.1 - 5 - 6 569 63
AK314779.1 - 4 - 5 498 63
AL514316.3 - 4 - 5 505 63
BC013138.1 - 4 - 5 483 4454722 63
BT006672.1 - 3 - 4 427 63
DC350361.1 - 4 - 5 535 63
L09235.1 - 3 - 4 442 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008369.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000583.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP6V1A

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
ATP metabolic process
phagosome acidification
ATP binding
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03010622.hg.1 ENST00000273398.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010622.hg.1 ENST00000538620.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010622.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010622.hg.1 TC03000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000462855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000462855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000462855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay PSR0300159755.hg.1 TC0300008369.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000273398.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000461496.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000462855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000462855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000462855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000470455.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000475322.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 ENST00000496747.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq
Affymetrix Human Clariom D Assay TC0300008369.hg.1 NM_001690.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0