This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01095225_s1
See other MFSD12 GE Assays ›
Gene Symbol:
MFSD12
Gene Name:
major facilitator superfamily domain containing 12
Gene Aliases:
C19orf28, PP3501
Chromosome Location:
Chr.19: 3538270 - 3557592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01095225_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MFSD12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF218008.1 - 1 1239 152
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG741167.1 - 1 414 Chr. 19 - 3538492 - 3539219 [-] on Build GRCh38 152
GenBankmRNA BM981481.1 - 1 Chr. 19 - 3538271 - 3538933 [+] on Build GRCh38 151
GenBankmRNA HY043798.1 - 1 255 Chr. 19 - 3538606 - 3539059 [-] on Build GRCh38 152
GenBankmRNA BU543927.1 - 1 294 Chr. 19 - 3538289 - 3539097 [-] on Build GRCh38 152
GenBankmRNA CB995863.1 - 160 Chr. - - [-] on Build 152

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream deoxyhypusine hydroxylase/monooxygenase DOHH HLRC1,
hDOHH
-32.4kb Sense Chr. 19 - 3490821 - 3500940 on NCBI Build GRCh38
Downstream chromosome 19 open reading frame 71 C19orf71 60.0 Anti-sense Chr. 19 - 3539157 - 3544030 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0