This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091750_m1
See other PPIH GE Assays ›
Gene Symbol:
PPIH
Gene Name:
peptidylprolyl isomerase H
Gene Aliases:
CYP-20, CYPH, SnuCyp-20, USA-CYP
Chromosome Location:
Chr.1: 42657770 - 42681654 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091750_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011540508.1 XP_011538810.1 4 - 5 342 115
XM_017000058.1 XP_016855547.1 4 - 5 996 115
XM_017000059.1 XP_016855548.1 4 - 5 342 115
GenBank mRNA AK294288.1 - 2 - 3 499 115
AK309438.1 - 4 - 5 338 115

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK294288.1,XM_011540508.1,XM_017000058.1,XM_017000059.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
protein peptidyl-prolyl isomerization
protein folding
protein complex assembly
positive regulation of viral genome replication
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
ribonucleoprotein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0