This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091358_g1
See other ATP6V1E1 GE Assays ›
Gene Symbol:
ATP6V1E1
Gene Name:
ATPase H+ transporting V1 subunit E1
Gene Aliases:
ATP6E, ATP6E2, ATP6V1E, P31, Vma4
Chromosome Location:
Chr.22: 17592136 - 17628822 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091358_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP6V1E1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039367.1 NP_001034456.1 4 - 5 462 108
GenBank mRNA AK315941.1 - 4 - 5 388 108
BE735148.1 - 4 - 5 408 108
BI597419.1 - 4 - 5 392 108

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
phagosome acidification
protein binding
hydrogen-exporting ATPase activity, phosphorylative mechanism
proton-transporting ATPase activity, rotational mechanism
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0