This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091240_m1
See other RNF17 GE Assays ›
Gene Symbol:
RNF17
Gene Name:
ring finger protein 17
Gene Aliases:
Mmip-2, SPATA23, TDRD4
Chromosome Location:
Chr.13: 24747641 - 24888816 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091240_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNF17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011535163.1 XP_011533465.1 1 - 2 91 96
GenBank mRNA AF285603.1 - 11 - 12 1391 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011535163.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatid development
zinc ion binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0