This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091210_g1
See other AMMECR1L GE Assays ›
Gene Symbol:
AMMECR1L
Gene Name:
AMMECR1 like
Gene Aliases:
-
Chromosome Location:
Chr.2: 127861630 - 127886408 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091210_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AMMECR1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199140.1 NP_001186069.1 7 - 8 1097 88
NM_031445.2 NP_113633.2 7 - 8 1069 88
XM_005263804.4 XP_005263861.1 8 - 9 1700 88
XM_005263806.4 XP_005263863.1 7 - 8 1589 88
XM_005263807.4 XP_005263864.1 6 - 7 1454 88
XM_011511952.2 XP_011510254.1 7 - 8 1590 88
XM_011511954.2 XP_011510256.1 8 - 9 1677 88
XM_011511955.2 XP_011510257.1 6 - 7 619 88
GenBank mRNA AK095871.1 - 2 - 3 154 88
AK304145.1 - 7 - 8 1097 88
BC016181.1 - 7 - 8 1069 4644159 88
BC078174.1 - 7 - 8 1079 6012672 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014190.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002289.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AMMECR1L

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036817.hg.1 ENST00000272647.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036817.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036817.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036817.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036817.hg.1 TC02002289.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203714.hg.1 TC0200014190.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203715.hg.1 TC0200014190.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0