This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091206_g1
See other AMMECR1L GE Assays ›
Gene Symbol:
AMMECR1L
Gene Name:
AMMECR1 like
Gene Aliases:
-
Chromosome Location:
Chr.2: 127861630 - 127886408 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091206_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AMMECR1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199140.1 NP_001186069.1 3 - 4 690 64
NM_031445.2 NP_113633.2 3 - 4 662 64
XM_005263804.4 XP_005263861.1 3 - 4 1158 64
XM_011511952.2 XP_011510254.1 2 - 3 1048 64
XM_011511956.2 XP_011510258.1 3 - 4 1158 64
XM_017005063.1 XP_016860552.1 3 - 4 1158 64
GenBank mRNA AK304145.1 - 3 - 4 690 64
BC016181.1 - 3 - 4 662 4644159 64
BC078174.1 - 3 - 4 672 6012672 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014190.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002289.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AMMECR1L

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02036821.hg.1 ENST00000272647.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036821.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036821.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036821.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036821.hg.1 TC02002289.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay PSR0200203722.hg.1 TC0200014190.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000272647.9 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 ENST00000393001.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_001199140.1 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq
Affymetrix Human Clariom D Assay TC0200014190.hg.1 NM_031445.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0