This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01089791_m1
See other CREBL2 GE Assays ›
Gene Symbol:
CREBL2
Gene Name:
cAMP responsive element binding protein like 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 12611833 - 12645108 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01089791_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CREBL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001310.3 NP_001301.1 2 - 3 557 104
GenBank mRNA AB451461.1 - 1 - 2 217 104
AF039081.1 - 2 - 3 493 104
BC104873.1 - 2 - 3 401 8143876 104
BC106052.1 - 2 - 3 498 6150812 104
BC112157.1 - 2 - 3 401 8327625 104
BY795169.2 - 2 - 3 557 104
CR542250.1 - 1 - 2 217 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200006881.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000176.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CREBL2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cell cycle
signal transduction
cell differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of glucose import
positive regulation of lipid biosynthetic process
protein stabilization
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12002233.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12002233.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12002233.hg.1 TC12000176.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay PSR1200148103.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200148103.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay PSR1200148103.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200148103.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay PSR1200148103.hg.1 TC1200006881.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000228865.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 ENST00000540224.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq
Affymetrix Human Clariom D Assay TC1200006881.hg.1 NM_001310.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0