This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01089087_m1
See other CPSF3 GE Assays ›
Gene Symbol:
CPSF3
Gene Name:
cleavage and polyadenylation specific factor 3
Gene Aliases:
CPSF-73, CPSF73
Chromosome Location:
Chr.2: 9423537 - 9473110 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01089087_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPSF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321833.1 NP_001308762.1 3 - 4 679 106
NM_001321834.1 NP_001308763.1 3 - 4 1587 106
NM_001321835.1 NP_001308764.1 3 - 4 450 106
NM_001321836.1 NP_001308765.1 4 - 5 769 106
NM_016207.3 NP_057291.1 3 - 4 450 106
GenBank mRNA AF171877.1 - 3 - 4 228 106
AK025440.1 - 1 - 2 109 106
AK223487.1 - 3 - 4 248 106
AK298918.1 - 3 - 4 326 106
BC011654.2 - 3 - 4 323 4304135 106
BC014106.1 - 3 - 4 1240 106
BC020211.1 - 3 - 4 248 4890593 106
BC030988.1 - 3 - 4 336 106
BG773037.1 - 4 - 5 322 106
DA478774.1 - 3 - 4 326 106
DC420903.1 - 3 - 4 450 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200006647.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK298918.1,NM_001321835.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CPSF3

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA polyadenylation
mRNA cleavage
mRNA 3'-end processing by stem-loop binding and cleavage
mRNA export from nucleus
mRNA 3'-end processing
RNA phosphodiester bond hydrolysis, endonucleolytic
RNA binding
endoribonuclease activity
protein binding
5'-3' exonuclease activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489403.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 ENST00000489629.1 Ensembl
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay TC0200006647.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 ENST00000238112.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 ENST00000460593.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 ENST00000475482.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 NM_016207.3 Refseq
Affymetrix Human Clariom D Assay PSR0200145467.hg.1 TC0200006647.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0