This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01086901_m1
See other TMEFF2 GE Assays ›
Gene Symbol:
TMEFF2
Gene Name:
transmembrane protein with EGF like and two follistatin like domains 2
Gene Aliases:
CT120.2, HPP1, TENB2, TPEF, TR, TR-2
Chromosome Location:
Chr.2: 191948299 - 192194940 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01086901_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEFF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001305134.1 NP_001292063.1 1 - 2 577 85
NM_001305145.1 NP_001292074.1 1 - 2 577 85
NM_016192.3 NP_057276.2 1 - 2 577 85
XM_011510890.2 XP_011509192.1 1 - 2 572 85
XM_017003739.1 XP_016859228.1 1 - 2 584 85
XM_017003740.1 XP_016859229.1 1 - 2 584 85
GenBank mRNA AB004064.1 - 246 85
AB017269.1 - 1 - 2 535 85
AF179274.2 - 1 - 2 562 85
AF242222.1 - 1 - 2 577 85
AK054646.1 - 1 - 2 1357 85
AK074507.1 - 1 - 2 191 85
AK074632.1 - 1 - 2 560 85
AK222691.1 - 1 - 2 582 85
AK315376.1 - 1 - 2 578 85
AL157430.1 - 1 - 2 560 85
AY358907.1 - 1 - 2 543 85
BC008973.1 - 1 - 2 518 4180709 85
CR457390.1 - 1 - 2 168 85
DQ133599.1 - 1 - 2 168 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200015266.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002628.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMEFF2

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell migration
wound healing, spreading of cells
negative regulation of integrin biosynthetic process
negative regulation of stress fiber assembly
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000272771.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000392314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000409056.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000409056.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000409056.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000409056.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000409056.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000487771.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000487771.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 ENST00000487771.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305134.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_001305145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200015266.hg.1 NM_016192.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 216382 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0