Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014167.hg.1 |
ENST00000427522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014167.hg.1 |
ENST00000467148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014167.hg.1 |
NM_001037335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014167.hg.1 |
NM_033405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014167.hg.1 |
TC20001035.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
ENST00000427522.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
ENST00000467148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
ENST00000478861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
NM_001037335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
NM_033405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
ENST00000427522.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
ENST00000467148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
ENST00000478861.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
NM_001037335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
NM_033405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168557.hg.1 |
TC2000009819.hg.1 |
Clariom D TC |