This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01085999_m1
See other TGFB3 GE Assays ›
Gene Symbol:
TGFB3
Gene Name:
transforming growth factor beta 3
Gene Aliases:
ARVD, ARVD1, LDS5, RNHF, TGF-beta3
Chromosome Location:
Chr.14: 75958097 - 75982022 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01085999_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGFB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003239.3 NP_003230.1 5 - 6 2055 65
GenBank mRNA AK122902.1 - 2 - 3 1307 65
AK313564.1 - 5 - 6 1136 65
EF560714.1 - 5 - 6 1039 65
J03241.1 - 5 - 6 1188 65
X14149.1 - 5 - 6 1179 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400009738.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TGFB3

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
response to hypoxia
in utero embryonic development
platelet degranulation
transforming growth factor beta receptor signaling pathway
SMAD protein import into nucleus
salivary gland morphogenesis
female pregnancy
aging
negative regulation of DNA replication
negative regulation of cell proliferation
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of macrophage cytokine production
cell growth
positive regulation of bone mineralization
negative regulation of transforming growth factor beta receptor signaling pathway
mammary gland development
response to progesterone
positive regulation of collagen biosynthetic process
response to laminar fluid shear stress
wound healing
regulation of cell proliferation
odontogenesis
uterine wall breakdown
regulation of apoptotic process
positive regulation of apoptotic process
regulation of MAPK cascade
negative regulation of neuron apoptotic process
response to estrogen
ossification involved in bone remodeling
cell-cell junction organization
positive regulation of DNA replication
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lung alveolus development
digestive tract development
embryonic neurocranium morphogenesis
inner ear development
positive regulation of protein secretion
positive regulation of filopodium assembly
positive regulation of stress fiber assembly
positive regulation of cell division
palate development
face morphogenesis
frontal suture morphogenesis
positive regulation of SMAD protein import into nucleus
SMAD protein signal transduction
detection of hypoxia
regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of occluding junction disassembly
type II transforming growth factor beta receptor binding
cytokine activity
protein binding
growth factor activity
type I transforming growth factor beta receptor binding
type III transforming growth factor beta receptor binding
identical protein binding
protein heterodimerization activity
transforming growth factor beta binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556507.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556507.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556507.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay PSR1400171868.hg.1 TC1400009738.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556507.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556507.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556507.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0