This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01085355_m1
See other STK39 GE Assays ›
Gene Symbol:
STK39
Gene Name:
serine/threonine kinase 39
Gene Aliases:
DCHT, PASK, SPAK
Chromosome Location:
Chr.2: 167954020 - 168247595 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01085355_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STK39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013233.2 NP_037365.2 4 - 5 731 136
XM_005246465.2 XP_005246522.1 4 - 5 677 136
XM_017003813.1 XP_016859302.1 4 - 5 677 136
XM_017003814.1 XP_016859303.1 4 - 5 677 136
XM_017003815.1 XP_016859304.1 4 - 5 2017 136
XM_017003816.1 XP_016859305.1 4 - 5 677 136
GenBank mRNA AF099989.1 - 4 - 5 750 136
KJ534948.1 - 5 - 6 514 136
KJ534949.1 - 5 - 6 514 136
KJ534950.1 - 5 - 6 526 136
KJ534951.1 - 5 - 6 505 136
KJ535059.1 - 5 - 6 494 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014834.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002504.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STK39

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
protein phosphorylation
response to stress
regulation of blood pressure
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
intracellular signal transduction
positive regulation of potassium ion transport
regulation of inflammatory response
cellular hypotonic response
negative regulation of pancreatic juice secretion
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
regulation of ion homeostasis
negative regulation of rubidium ion transport
negative regulation of rubidium ion transmembrane transporter activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000461000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000461000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000461000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02039653.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02039653.hg.1 NM_013233.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02039653.hg.1 TC02002504.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000461000.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000461000.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000461000.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 ENST00000487143.5 Ensembl
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay TC0200014834.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay PSR0200208239.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200208239.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay PSR0200208239.hg.1 ENST00000355999.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200208239.hg.1 NM_013233.2 Refseq
Affymetrix Human Clariom D Assay PSR0200208239.hg.1 TC0200014834.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0