This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01085340_m1
See other ACE2 GE Assays ›
Gene Symbol:
ACE2
Gene Name:
angiotensin I converting enzyme 2
Gene Aliases:
ACEH
Chromosome Location:
Chr.X: 15494525 - 15602069 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01085340_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021804.2 NP_068576.1 7 - 8 1017 122
XM_011545549.1 XP_011543851.1 6 - 7 854 122
XM_011545551.2 XP_011543853.1 6 - 7 854 122
XM_011545552.1 XP_011543854.1 6 - 7 854 122
GenBank mRNA AB046569.1 - 6 - 7 854 122
AB193259.1 - 7 - 8 1103 122
AB193260.1 - 6 - 7 968 122
AF241254.1 - 6 - 7 903 122
AF291820.1 - 6 - 7 820 122
AK315144.1 - 7 - 8 1018 122
AL110224.1 - 6 - 7 799 122
AY358714.1 - 6 - 7 839 122
AY623811.1 - 6 - 7 800 122
BC039902.1 - 6 - 7 818 5243048 122
BC048094.1 - 6 - 7 834 5297380 122
BC048144.1 - 6 - 7 834 122
GQ262784.1 - 6 - 7 800 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009128.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000878.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ACE2

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

regulation of cytokine production
angiotensin maturation
angiotensin catabolic process in blood
angiotensin-mediated drinking behavior
regulation of systemic arterial blood pressure by renin-angiotensin
tryptophan transport
regulation of vasoconstriction
receptor biosynthetic process
regulation of cell proliferation
regulation of vasodilation
viral entry into host cell
receptor-mediated virion attachment to host cell
regulation of inflammatory response
positive regulation of cardiac muscle contraction
positive regulation of gap junction assembly
regulation of cardiac conduction
positive regulation of reactive oxygen species metabolic process
virus receptor activity
glycoprotein binding
endopeptidase activity
carboxypeptidase activity
metallocarboxypeptidase activity
protein binding
metallopeptidase activity
peptidyl-dipeptidase activity
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X011761.hg.1 ENST00000252519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X011761.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X011761.hg.1 NM_021804.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X011761.hg.1 TC0X000878.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000471548.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000473851.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 ENST00000484756.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009128.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 ENST00000252519.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 ENST00000427411.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 NM_021804.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00162650.hg.1 TC0X00009128.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0