Assay Details
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153002.2 | NP_694547.2 | 5 - 6 | 798 | 61 | |
XM_005247165.2 | XP_005247222.1 | 6 - 7 | 1075 | 61 | ||
XM_011512484.2 | XP_011510786.1 | 7 - 8 | 1148 | 61 | ||
XM_011512487.1 | XP_011510789.1 | 5 - 6 | 700 | 61 | ||
XM_017005795.1 | XP_016861284.1 | 6 - 7 | 1111 | 61 | ||
XM_017005796.1 | XP_016861285.1 | 6 - 7 | 1120 | 61 | ||
GenBank mRNA | AF488739.1 | - | 5 - 6 | 601 | 61 | |
AY255565.1 | - | 2 - 3 | 190 | 61 | ||
BC113701.1 | - | 5 - 6 | 621 | 8322753 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001689.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03030468.hg.1 | ENST00000315843.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03030468.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03030468.hg.1 | ENST00000464295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03030468.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03030468.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03030468.hg.1 | TC03001689.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300190336.hg.1 | TC0300012140.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000315843.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000461057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000464295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000481907.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | ENST00000495912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_001168271.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012140.hg.1 | NM_153002.2 | Refseq |
