This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01083793_m1
See other GPR156 GE Assays ›
Gene Symbol:
GPR156
Gene Name:
G protein-coupled receptor 156
Gene Aliases:
GABABL, PGR28
Chromosome Location:
Chr.3: 120165481 - 120285122 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01083793_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR156
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168271.1 NP_001161743.1 4 - 5 665 68
NM_153002.2 NP_694547.2 4 - 5 665 68
XM_005247165.2 XP_005247222.1 5 - 6 942 68
XM_011512484.2 XP_011510786.1 6 - 7 1015 68
XM_011512487.1 XP_011510789.1 4 - 5 567 68
XM_017005795.1 XP_016861284.1 5 - 6 978 68
XM_017005796.1 XP_016861285.1 5 - 6 987 68
XM_017005797.1 XP_016861286.1 5 - 6 942 68
GenBank mRNA AF488739.1 - 4 - 5 468 68
AY255565.1 - 1 - 2 57 68
BC113701.1 - 4 - 5 488 8322753 68
BC143606.1 - 4 - 5 488 9052119 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012140.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001689.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR156

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled GABA receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030470.hg.1 ENST00000315843.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030470.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030470.hg.1 ENST00000464295.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030470.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030470.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030470.hg.1 TC03001689.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190338.hg.1 TC0300012140.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0