This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01083792_g1
See other GPR156 GE Assays ›
Gene Symbol:
GPR156
Gene Name:
G protein-coupled receptor 156
Gene Aliases:
GABABL, PGR28
Chromosome Location:
Chr.3: 120165481 - 120285122 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01083792_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR156
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168271.1 NP_001161743.1 1 - 2 284 93
NM_153002.2 NP_694547.2 1 - 2 284 93
XM_005247165.2 XP_005247222.1 2 - 3 561 93
XM_011512484.2 XP_011510786.1 2 - 3 561 93
XM_017005795.1 XP_016861284.1 2 - 3 597 93
XM_017005796.1 XP_016861285.1 2 - 3 606 93
XM_017005797.1 XP_016861286.1 2 - 3 561 93
GenBank mRNA AB209362.1 - 1 - 2 154 93
AF488739.1 - 1 - 2 87 93
AK310204.1 - 2 - 3 513 93
BC113701.1 - 1 - 2 107 8322753 93
BC143606.1 - 1 - 2 107 9052119 93
CD638213.1 - 2 - 3 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012140.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001689.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_011512484.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR156

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled GABA receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03030475.hg.1 ENST00000315843.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030475.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030475.hg.1 ENST00000464295.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030475.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030475.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030475.hg.1 TC03001689.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03030474.hg.1 ENST00000315843.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030474.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030474.hg.1 ENST00000464295.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030474.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030474.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030474.hg.1 TC03001689.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030477.hg.1 ENST00000315843.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030477.hg.1 ENST00000464295.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030477.hg.1 NM_001168271.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030477.hg.1 NM_153002.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030477.hg.1 TC03001689.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190342.hg.1 TC0300012140.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190346.hg.1 TC0300012140.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay TC0300012140.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000315843.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000461057.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000464295.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000481907.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 ENST00000495912.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 NM_001168271.1 Refseq
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 NM_153002.2 Refseq
Affymetrix Human Clariom D Assay PSR0300190343.hg.1 TC0300012140.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0