This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01083130_m1
See other PPIP5K2 GE Assays ›
Gene Symbol:
PPIP5K2
Gene Name:
diphosphoinositol pentakisphosphate kinase 2
Gene Aliases:
CFAP160, HISPPD1, IP7K2, VIP2
Chromosome Location:
Chr.5: 103120254 - 103203523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01083130_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIP5K2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015216.3 NP_056031.2 26 - 27 3575 92
XM_005271935.1 XP_005271992.1 27 - 28 3809 92
XM_005277536.1 XP_005277593.1 28 - 29 3854 92
XM_017009272.1 XP_016864761.1 26 - 27 3635 92
XM_017009273.1 XP_016864762.1 26 - 27 3632 92
GenBank mRNA BC024591.1 - 26 - 27 3328 3913521 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500008232.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000498.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPIP5K2

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

inositol metabolic process
phosphorylation
dephosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
acid phosphatase activity
ATP binding
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05007032.hg.1 ENST00000321521.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007032.hg.1 ENST00000358359.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007032.hg.1 ENST00000414217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007032.hg.1 ENST00000451606.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007032.hg.1 NM_015216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05007032.hg.1 TC05000498.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000321521.13 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000358359.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000414217.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000509597.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000510672.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000511022.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000613674.4 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000613907.4 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000627916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 NM_001276277.2 Refseq
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 NM_001281471.2 Refseq
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 NM_015216.4 Refseq
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000321521.13 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000358359.7 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000414217.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000509597.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000510672.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000511022.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000613674.4 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000613907.4 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 ENST00000627916.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 NM_001276277.2 Refseq
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 NM_001281471.2 Refseq
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 NM_015216.4 Refseq
Affymetrix Human Clariom D Assay PSR0500156418.hg.1 TC0500008232.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0