This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01082921_m1
See other FAT1 GE Assays ›
Gene Symbol:
FAT1
Gene Name:
FAT atypical cadherin 1
Gene Aliases:
CDHF7, CDHR8, FAT, ME5, hFat1
Chromosome Location:
Chr.4: 186587783 - 186726915 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01082921_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005245.3 NP_005236.2 26 - 27 13329 73
XM_006714139.2 XP_006714202.1 26 - 27 13721 73
GenBank mRNA X87241.1 - 26 - 27 13333 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400012670.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001824.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FAT1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
establishment or maintenance of cell polarity
cell-cell signaling
anatomical structure morphogenesis
single organismal cell-cell adhesion
cell migration
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000441802.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000500085.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000503253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000503253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000503253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000507662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000508035.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000508035.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000508035.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509537.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000509927.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512347.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000512772.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 ENST00000614102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012670.hg.1 NM_005245.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 131558 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0