This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01082139_m1
See other PFKP GE Assays ›
Gene Symbol:
PFKP
Gene Name:
phosphofructokinase, platelet
Gene Aliases:
ATP-PFK, PFK-C, PFK-P, PFKF
Chromosome Location:
Chr.10: 3067475 - 3136805 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01082139_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PFKP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323068.1 NP_001309997.1 1 - 2 184 134
NM_001323069.1 NP_001309998.1 1 - 2 184 134
NM_002627.4 NP_002618.1 1 - 2 184 134
XM_005252465.4 XP_005252522.1 1 - 2 229 134
XM_005252466.4 XP_005252523.1 1 - 2 229 134
GenBank mRNA AK291841.1 - 1 - 2 156 134
BC002536.1 - 1 - 2 144 3140233 134
BC029138.1 - 1 - 2 184 5180268 134
D25328.1 - 1 - 2 141 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012423.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000022.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001323069.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PFKP

Panther Classification:

Molecular Function -

carbohydrate kinase kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

fructose 6-phosphate metabolic process
canonical glycolysis
cell-cell adhesion
mitophagy in response to mitochondrial depolarization
6-phosphofructokinase activity
ATP binding
protein complex binding
metal ion binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381072.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000381125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000407806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000413079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000413079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000413079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000413079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000413079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000415005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000421751.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000421751.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000421751.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000421751.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000421751.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000421751.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000433193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000433193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000433193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000433193.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000460445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000460445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000460445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000460445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000460445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000468050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000495715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000495715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000607886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000607886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000607886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 ENST00000607886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_001242339.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012423.hg.1 NM_002627.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 128305 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0