Assay Details
Gene Symbol: CCBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133459.3 | NP_597716.1 | 3 - 4 | 330 | 94 | |
XM_017025556.1 | XP_016881045.1 | 3 - 4 | 1097 | 94 | ||
XM_017025557.1 | XP_016881046.1 | 4 - 5 | 990 | 94 | ||
XM_017025558.1 | XP_016881047.1 | 3 - 4 | 1098 | 94 | ||
GenBank mRNA | AB075863.1 | - | 3 - 4 | 298 | 94 | |
AY358347.1 | - | 3 - 4 | 276 | 94 | ||
BF434122.1 | - | 3 - 4 | 330 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000548.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lymphangiogenesis
sprouting angiogenesis
positive regulation of vascular endothelial growth factor production
positive regulation of endothelial cell migration
positive regulation of protein processing
positive regulation of angiogenesis
venous blood vessel morphogenesis
positive regulation of vascular endothelial growth factor signaling pathway
positive regulation of lymphangiogenesis
protease binding
calcium ion binding
protein binding
collagen binding
lymphangiogenesis
sprouting angiogenesis
positive regulation of vascular endothelial growth factor production
positive regulation of endothelial cell migration
positive regulation of protein processing
positive regulation of angiogenesis
venous blood vessel morphogenesis
positive regulation of vascular endothelial growth factor signaling pathway
positive regulation of lymphangiogenesis
protease binding
calcium ion binding
protein binding
collagen binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005698.hg.1 | ENST00000398179.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005698.hg.1 | ENST00000439986.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005698.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005698.hg.1 | TC18000548.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800160793.hg.1 | TC1800008827.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000398179.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000439986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589116.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | ENST00000589419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008827.hg.1 | NM_133459.3 | Refseq |
