Assay Details
Gene Symbol: NACC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144653.4 | NP_653254.1 | 3 - 4 | 1206 | 86 | |
XM_011518221.2 | XP_011516523.1 | 3 - 4 | 1226 | 86 | ||
GenBank mRNA | BC015649.2 | - | 3 - 4 | 1191 | 4654320 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001722.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of cell proliferation
negative regulation of cell proliferation
posttranscriptional regulation of gene expression
histone deacetylation
cellular protein complex localization
negative regulation of transcription, DNA-templated
protein homooligomerization
negative regulation of G1/S transition of mitotic cell cycle by negative regulation of transcription from RNA polymerase II promoter
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
histone deacetylase activity
protein binding
protein homodimerization activity
histone deacetylase binding
transcription, DNA-templated
positive regulation of cell proliferation
negative regulation of cell proliferation
posttranscriptional regulation of gene expression
histone deacetylation
cellular protein complex localization
negative regulation of transcription, DNA-templated
protein homooligomerization
negative regulation of G1/S transition of mitotic cell cycle by negative regulation of transcription from RNA polymerase II promoter
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
histone deacetylase activity
protein binding
protein homodimerization activity
histone deacetylase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09022003.hg.1 | ENST00000277554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09022003.hg.1 | ENST00000371753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09022003.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09022003.hg.1 | TC09001722.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09022005.hg.1 | ENST00000277554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09022005.hg.1 | ENST00000371753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09022005.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09022005.hg.1 | TC09001722.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000467669.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | ENST00000467669.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09022004.hg.1 | ENST00000277554.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09022004.hg.1 | ENST00000371753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09022004.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09022004.hg.1 | TC09001722.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181795.hg.1 | TC0900011962.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181796.hg.1 | TC0900011962.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000467669.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | ENST00000467669.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011962.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | ENST00000277554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | ENST00000371753.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | NM_144653.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900181797.hg.1 | TC0900011962.hg.1 | Clariom D TC |
