This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01074088_m1
See other ABCC9 GE Assays ›
Gene Symbol:
ABCC9
Gene Name:
ATP binding cassette subfamily C member 9
Gene Aliases:
ABC37, ATFB12, CANTU, CMD1O, SUR2
Chromosome Location:
Chr.12: 21797389 - 21941869 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01074088_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCC9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020297.3 NP_064693.2 37 - 38 4534 129
XM_005253284.3 XP_005253341.1 39 - 40 5320 129
XM_005253286.3 XP_005253343.1 40 - 41 5380 129
XM_005253288.3 XP_005253345.1 38 - 39 5204 129
XM_005253289.3 XP_005253346.1 38 - 39 5281 129
XM_005253290.3 XP_005253347.1 37 - 38 5179 129
XM_011520545.2 XP_011518847.1 39 - 40 5376 129
GenBank mRNA AK092535.1 - 13 - 14 1353 129
AK094171.1 - 4 - 5 381 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200010111.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001300.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABCC9

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
signal transduction
potassium ion import
defense response to virus
transmembrane transport
anion transmembrane transport
regulation of cardiac conduction
transporter activity
potassium channel activity
ATP binding
sulfonylurea receptor activity
potassium channel regulator activity
anion transmembrane-transporting ATPase activity
ion channel binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12016987.hg.1 ENST00000261200.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12016987.hg.1 NM_020297.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12016987.hg.1 TC12001300.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200176418.hg.1 ENST00000261200.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200176418.hg.1 NM_020297.3 Refseq
Affymetrix Human Clariom D Assay PSR1200176418.hg.1 ENST00000261200.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200176418.hg.1 NM_020297.3 Refseq
Affymetrix Human Clariom D Assay PSR1200176418.hg.1 TC1200010111.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0