This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01073732_m1
See other EPB41L4B GE Assays ›
Gene Symbol:
EPB41L4B
Gene Name:
erythrocyte membrane protein band 4.1 like 4B
Gene Aliases:
CG1, EHM2
Chromosome Location:
Chr.9: 109171974 - 109320964 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01073732_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41L4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018424.3 NP_060894.2 14 - 15 1865 77
NM_019114.4 NP_061987.3 14 - 15 1865 77
XM_011518790.1 XP_011517092.1 14 - 15 1362 77
XM_011518791.2 XP_011517093.1 14 - 15 1347 77
XM_011518792.2 XP_011517094.1 14 - 15 1347 77
XM_011518794.2 XP_011517096.1 14 - 15 1347 77
XM_017014813.1 XP_016870302.1 14 - 15 1347 77
XM_017014814.1 XP_016870303.1 14 - 15 1347 77
XM_017014815.1 XP_016870304.1 14 - 15 1347 77
GenBank mRNA AB032179.3 - 14 - 15 1865 77
AB209187.1 - 6 - 7 457 77
AK000388.1 - 14 - 15 1249 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011160.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EPB41L4B

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of gene expression
regulation of keratinocyte proliferation
actomyosin structure organization
wound healing
positive regulation of cell adhesion
positive regulation of keratinocyte migration
structural constituent of cytoskeleton
cytoskeletal protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 ENST00000374557.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 ENST00000374566.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 NM_018424.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 NM_019114.4 Refseq
Affymetrix Human Clariom D Assay PSR0900175917.hg.1 TC0900011160.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0