This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01071772_m1
See other XDH GE Assays ›
Gene Symbol:
XDH
Gene Name:
xanthine dehydrogenase
Gene Aliases:
XO, XOR
Chromosome Location:
Chr.2: 31334320 - 31414777 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01071772_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XDH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000379.3 NP_000370.2 9 - 10 875 114
XM_011533095.2 XP_011531397.1 9 - 10 907 114
XM_011533096.2 XP_011531398.1 9 - 10 906 114
GenBank mRNA D11456.2 - 9 - 10 853 114
U06117.1 - 9 - 10 859 114
U39487.1 - 9 - 10 830 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012163.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001714.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene XDH

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
purine nucleotide catabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
lactation
xanthine catabolic process
negative regulation of gene expression
negative regulation of endothelial cell differentiation
negative regulation of protein kinase B signaling
oxidation-reduction process
positive regulation of p38MAPK cascade
negative regulation of vascular endothelial growth factor signaling pathway
positive regulation of reactive oxygen species metabolic process
negative regulation of vasculogenesis
xanthine dehydrogenase activity
xanthine oxidase activity
iron ion binding
electron carrier activity
oxidoreductase activity, acting on CH-OH group of donors
oxidoreductase activity, acting on the aldehyde or oxo group of donors
protein homodimerization activity
molybdopterin cofactor binding
flavin adenine dinucleotide binding
2 iron, 2 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02027657.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02027657.hg.1 ENST00000491727.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02027657.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02027657.hg.1 TC02001714.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay PSR0200188800.hg.1 TC0200012163.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000379416.3 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000476043.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 ENST00000491727.5 Ensembl
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq
Affymetrix Human Clariom D Assay TC0200012163.hg.1 NM_000379.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0